missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RASA3 Full-length ORF (NP_031394.2, 1 a.a. - 834 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00022821-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16196612

  • 6245.00 NOK / 25µg
Estimated Shipment: 19-09-2024
to see stock.

Description

Description

The protein encoded by this gene is member of the GAP1 family of GTPase-activating proteins. The gene product stimulates the GTPase activity of normal RAS p21 but not its oncogenic counterpart. Acting as a suppressor of RAS function, the protein enhances the weak intrinsic GTPase activity of RAS proteins resulting in the inactive GDP-bound form of RAS, thereby allowing control of cellular proliferation and differentiation. This family member is an inositol 1,3,4,5-tetrakisphosphate-binding protein, like the closely related RAS p21 protein activator 2. The two family members have distinct pleckstrin-homology domains, with this particular member having a domain consistent with its localization to the plasma membrane. [provided by RefSeq]

Sequence: MAVEDEGLRVFQSVKIKIGEAKNLPSYPGPSKMRDCYCTVNLDQEEVFRTKIVEKSLCPFYGEDFYCEIPRSFRHLSFYIFDRDVFRRDSIIGKVAIQKEDLQKYHNRDTWFQLQHVDADSEVQGKVHLELRLSEVITDTGVVCHKLATRIVECQGLPIVNGQCDPYATVTLAGPFRSEAKKTKVKRKTNNPQFDEVFYFEVTRPCSYSKKSHFDFEEEDVDKLEIRVDLWNASNLKFGDEFLGELRIPLKVLRQSSSYEAWYFLQPRDNGSKSLKPDDLGSLRLNVVYTEDHVFSSDYYSPLRDLLLKSADVEPVSASAAHILGEVCREKQEAAVPLVRLFLHYGRVVPFISAIASAEVKRTQDPNTIFRGNSLASKCIDETMKLAGMHYLHVTLKPAIEEICQSHKPCEIDPVKLKDGENLENNMENLRQYVDRVFHAITESGVSCPTVMCDIFFSLREAAAKRFQDDPDVRYTAVSSFIFLRFFAPAILSPNLFQLTPHHTDPQTSRTLTLISKTVQTLGSLSKSKSASFKESYMATFYEFFNEQKYADAVKNFLDLISSSGRRDPKSVEQPIVLKEGFMIKRAQGRKRFGMKNFKKRWFRLTNHEFTYHKSKGDQPLYSIPIENILAVEKLEEESFKMKNMFQVIQPERALYIQANNCVEAKDWIDILTKVSQCNQKRLTVYHPSAYLSGHWLCCRAPSDSAPGCSPCTGGLPANIQLDIDGDRETERIYSLFNLYMSKLEKMQEACGSKSVYDGPEQEEYSTFVIDDPQETYKTLKQVIAGVGALEQEHAQYKRDKFKKTKYGSQEHPIGDKSFQNYIRQQSETSTHSI
Specifications

Specifications

NP_031394.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
122.1kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GAP1IP4BP/GAPIII/MGC46517/MGC47588
RASA3
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
22821
RASA3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAVEDEGLRVFQSVKIKIGEAKNLPSYPGPSKMRDCYCTVNLDQEEVFRTKIVEKSLCPFYGEDFYCEIPRSFRHLSFYIFDRDVFRRDSIIGKVAIQKEDLQKYHNRDTWFQLQHVDADSEVQGKVHLELRLSEVITDTGVVCHKLATRIVECQGLPIVNGQCDPYATVTLAGPFRSEAKKTKVKRKTNNPQFDEVFYFEVTRPCSYSKKSHFDFEEEDVDKLEIRVDLWNASNLKFGDEFLGELRIPLKVLRQSSSYEAWYFLQPRDNGSKSLKPDDLGSLRLNVVYTEDHVFSSDYYSPLRDLLLKSADVEPVSASAAHILGEVCREKQEAAVPLVRLFLHYGRVVPFISAIASAEVKRTQDPNTIFRGNSLASKCIDETMKLAGMHYLHVTLKPAIEEICQSHKPCEIDPVKLKDGENLENNMENLRQYVDRVFHAITESGVSCPTVMCDIFFSLREAAAKRFQDDPDVRYTAVSSFIFLRFFAPAILSPNLFQLTPHHTDPQTSRTLTLISKTVQTLGSLSKSKSASFKESYMATFYEFFNEQKYADAVKNFLDLISSSGRRDPKSVEQPIVLKEGFMIKRAQGRKRFGMKNFKKRWFRLTNHEFTYHKSKGDQPLYSIPIENILAVEKLEEESFKMKNMFQVIQPERALYIQANNCVEAKDWIDILTKVSQCNQKRLTVYHPSAYLSGHWLCCRAPSDSAPGCSPCTGGLPANIQLDIDGDRETERIYSLFNLYMSKLEKMQEACGSKSVYDGPEQEEYSTFVIDDPQETYKTLKQVIAGVGALEQEHAQYKRDKFKKTKYGSQEHPIGDKSFQNYIRQQSETSTHSI
RUO
RASA3
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers