Learn More
Abnova™ Human RASA3 Full-length ORF (NP_031394.2, 1 a.a. - 834 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00022821-P01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is member of the GAP1 family of GTPase-activating proteins. The gene product stimulates the GTPase activity of normal RAS p21 but not its oncogenic counterpart. Acting as a suppressor of RAS function, the protein enhances the weak intrinsic GTPase activity of RAS proteins resulting in the inactive GDP-bound form of RAS, thereby allowing control of cellular proliferation and differentiation. This family member is an inositol 1,3,4,5-tetrakisphosphate-binding protein, like the closely related RAS p21 protein activator 2. The two family members have distinct pleckstrin-homology domains, with this particular member having a domain consistent with its localization to the plasma membrane. [provided by RefSeq]
Sequence: MAVEDEGLRVFQSVKIKIGEAKNLPSYPGPSKMRDCYCTVNLDQEEVFRTKIVEKSLCPFYGEDFYCEIPRSFRHLSFYIFDRDVFRRDSIIGKVAIQKEDLQKYHNRDTWFQLQHVDADSEVQGKVHLELRLSEVITDTGVVCHKLATRIVECQGLPIVNGQCDPYATVTLAGPFRSEAKKTKVKRKTNNPQFDEVFYFEVTRPCSYSKKSHFDFEEEDVDKLEIRVDLWNASNLKFGDEFLGELRIPLKVLRQSSSYEAWYFLQPRDNGSKSLKPDDLGSLRLNVVYTEDHVFSSDYYSPLRDLLLKSADVEPVSASAAHILGEVCREKQEAAVPLVRLFLHYGRVVPFISAIASAEVKRTQDPNTIFRGNSLASKCIDETMKLAGMHYLHVTLKPAIEEICQSHKPCEIDPVKLKDGENLENNMENLRQYVDRVFHAITESGVSCPTVMCDIFFSLREAAAKRFQDDPDVRYTAVSSFIFLRFFAPAILSPNLFQLTPHHTDPQTSRTLTLISKTVQTLGSLSKSKSASFKESYMATFYEFFNEQKYADAVKNFLDLISSSGRRDPKSVEQPIVLKEGFMIKRAQGRKRFGMKNFKKRWFRLTNHEFTYHKSKGDQPLYSIPIENILAVEKLEEESFKMKNMFQVIQPERALYIQANNCVEAKDWIDILTKVSQCNQKRLTVYHPSAYLSGHWLCCRAPSDSAPGCSPCTGGLPANIQLDIDGDRETERIYSLFNLYMSKLEKMQEACGSKSVYDGPEQEEYSTFVIDDPQETYKTLKQVIAGVGALEQEHAQYKRDKFKKTKYGSQEHPIGDKSFQNYIRQQSETSTHSISpecifications
NP_031394.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
122.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GAP1IP4BP/GAPIII/MGC46517/MGC47588 | |
RASA3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
22821 | |
RASA3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAVEDEGLRVFQSVKIKIGEAKNLPSYPGPSKMRDCYCTVNLDQEEVFRTKIVEKSLCPFYGEDFYCEIPRSFRHLSFYIFDRDVFRRDSIIGKVAIQKEDLQKYHNRDTWFQLQHVDADSEVQGKVHLELRLSEVITDTGVVCHKLATRIVECQGLPIVNGQCDPYATVTLAGPFRSEAKKTKVKRKTNNPQFDEVFYFEVTRPCSYSKKSHFDFEEEDVDKLEIRVDLWNASNLKFGDEFLGELRIPLKVLRQSSSYEAWYFLQPRDNGSKSLKPDDLGSLRLNVVYTEDHVFSSDYYSPLRDLLLKSADVEPVSASAAHILGEVCREKQEAAVPLVRLFLHYGRVVPFISAIASAEVKRTQDPNTIFRGNSLASKCIDETMKLAGMHYLHVTLKPAIEEICQSHKPCEIDPVKLKDGENLENNMENLRQYVDRVFHAITESGVSCPTVMCDIFFSLREAAAKRFQDDPDVRYTAVSSFIFLRFFAPAILSPNLFQLTPHHTDPQTSRTLTLISKTVQTLGSLSKSKSASFKESYMATFYEFFNEQKYADAVKNFLDLISSSGRRDPKSVEQPIVLKEGFMIKRAQGRKRFGMKNFKKRWFRLTNHEFTYHKSKGDQPLYSIPIENILAVEKLEEESFKMKNMFQVIQPERALYIQANNCVEAKDWIDILTKVSQCNQKRLTVYHPSAYLSGHWLCCRAPSDSAPGCSPCTGGLPANIQLDIDGDRETERIYSLFNLYMSKLEKMQEACGSKSVYDGPEQEEYSTFVIDDPQETYKTLKQVIAGVGALEQEHAQYKRDKFKKTKYGSQEHPIGDKSFQNYIRQQSETSTHSI | |
RUO | |
RASA3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |