Learn More
Abnova™ Human RARRES1 Full-length ORF (NP_996846.1, 1 a.a. - 294 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005918-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene was identified as a retinoid acid (RA) receptor-responsive gene. It encodes a type 1 membrane protein. The expression of this gene is upregulated by tazarotene as well as by retinoic acid receptors. The expression of this gene is found to be downregulated in prostate cancer, which is caused by the methylation of its promoter and CpG island. Alternatively spliced transcript variant encoding distinct isoforms have been observed. [provided by RefSeq]
Sequence: MQPRRQRLPAPWSGPRGPRPTAPLLALLLLLAPVAAPAGSGDPDDPGQPQDAGVPRRLLQQAARAALHFFNFRSGSPSALRVLAEVQEGRAWINPKEGCKVHVVFSTERYNPESLLQEGEGRLGKCSARVFFKNQKPRPTINVTCTRLIEKKKRQQEDYLLYKQMKQLKNPLEIVSIPDNHGHIDPSLRLIWDLAFLGSSYVMWEMTTQVSHYYLAQLTSVRQWKTNDDTIDFDYTVLLHELSTQEIIPCRIHLVWYPGKPLKVKYHCQELQTPEEASGTEEGSAVVPTELSNFSpecifications
NP_996846.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
59.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TIG1 | |
RARRES1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
5918 | |
RARRES1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MQPRRQRLPAPWSGPRGPRPTAPLLALLLLLAPVAAPAGSGDPDDPGQPQDAGVPRRLLQQAARAALHFFNFRSGSPSALRVLAEVQEGRAWINPKEGCKVHVVFSTERYNPESLLQEGEGRLGKCSARVFFKNQKPRPTINVTCTRLIEKKKRQQEDYLLYKQMKQLKNPLEIVSIPDNHGHIDPSLRLIWDLAFLGSSYVMWEMTTQVSHYYLAQLTSVRQWKTNDDTIDFDYTVLLHELSTQEIIPCRIHLVWYPGKPLKVKYHCQELQTPEEASGTEEGSAVVPTELSNF | |
RUO | |
RARRES1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |