Learn More
Abnova™ Human RAPGEF2 Partial ORF (XP_376350, 1398 a.a. - 1487 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009693-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Members of the RAS (see HRAS; MIM 190020) subfamily of GTPases function in signal transduction as GTP/GDP-regulated switches that cycle between inactive GDP- and active GTP-bound states. Guanine nucleotide exchange factors (GEFs), such as RAPGEF2, serve as RAS activators by promoting acquisition of GTP to maintain the active GTP-bound state and are the key link between cell surface receptors and RAS activation (Rebhun et al., 2000 [PubMed 10934204]).[supplied by OMIM]
Sequence: PITDFPEGHSHPARKPPDYNVALQRSRMVARSSDTAGPSSVQQPHGHPTSSRPVNKPQWHKPNESDPRLAPYQSQGFSTEEDEDEQVSAVSpecifications
XP_376350 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.64kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PITDFPEGHSHPARKPPDYNVALQRSRMVARSSDTAGPSSVQQPHGHPTSSRPVNKPQWHKPNESDPRLAPYQSQGFSTEEDEDEQVSAV | |
RUO | |
RAPGEF2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9693 | |
RAPGEF2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CNrasGEF/NRAPGEP/PDZ-GEF1/PDZGEF1/RA-GEF/Rap-GEP | |
RAPGEF2 | |
Recombinant | |
wheat germ expression system |