Learn More
Abnova™ Human RAD54B Full-length ORF (AAH33710.2, 1 a.a. - 158 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | AAH33710.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 25788 |
Molecular Weight (g/mol) | 45kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16147732
|
Abnova™
H00025788-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16137732
|
Abnova™
H00025788-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene belongs to the DEAD-like helicase superfamily. It shares similarity with Saccharomyces cerevisiae RAD54 and RDH54, both of which are involved in homologous recombination and repair of DNA. This protein binds to double-stranded DNA, and displays ATPase activity in the presence of DNA. This gene is highly expressed in testis and spleen, which suggests active roles in meiotic and mitotic recombination. Homozygous mutations of this gene were observed in primary lymphoma and colon cancer. [provided by RefSeq]
Sequence: MLDHHPVAITVEVKQEEDIKPPPPLVLNSQQSDTLEQREEHELVHVMERSLSPSLSSVDMRMTSSPSSIPRRDDFFRHESGEHFRSLLGYDPQILQMLKEEHQIILENQKNFGLYVQEKRDGLKRRQQLEEELLRAKIEVEKLKAIRLRHDLPEYNSLSpecifications
AAH33710.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
45kDa | |
Glutathione Sepharose 4 Fast Flow | |
MLDHHPVAITVEVKQEEDIKPPPPLVLNSQQSDTLEQREEHELVHVMERSLSPSLSSVDMRMTSSPSSIPRRDDFFRHESGEHFRSLLGYDPQILQMLKEEHQIILENQKNFGLYVQEKRDGLKRRQQLEEELLRAKIEVEKLKAIRLRHDLPEYNSL | |
RUO | |
RAD54B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
25788 | |
RAD54B (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FSBP/RDH54 | |
RAD54B | |
Recombinant | |
wheat germ expression system |