missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RABGAP1L Partial ORF (NP_055672.3, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009910-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MEVRASLQKVSGSSDSVATMNSEEFVLVPQYADDNSTKHEEKPQLKIVSNGDEQLEKAMEEILRDSEKRPSSLLVDCQSSSEISDHSFGDIPASQTNKPSLQLILDPSNTSpecifications
NP_055672.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEVRASLQKVSGSSDSVATMNSEEFVLVPQYADDNSTKHEEKPQLKIVSNGDEQLEKAMEEILRDSEKRPSSLLVDCQSSSEISDHSFGDIPASQTNKPSLQLILDPSNT | |
RUO | |
RABGAP1L | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9910 | |
RABGAP1L (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686E1450/HHL/TBC1D18 | |
RABGAP1L | |
Recombinant | |
wheat germ expression system |