Learn More
Abnova™ Human RAB11FIP1 Full-length ORF (NP_001002233.1, 1 a.a. - 612 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00080223-P01.25ug
Additional Details : Weight : 0.00010kg
Description
Proteins of the large Rab GTPase family (see RAB1A; MIM 179508) have regulatory roles in the formation, targeting, and fusion of intracellular transport vesicles. RAB11FIP1 is one of many proteins that interact with and regulate Rab GTPases (Hales et al., 2001 [PubMed 11495908]).[supplied by OMIM]
Sequence: MGRTRETGTEKNTSSLELEESLPEQPETGRQEEELPRFPCKKQDYSPSSGEAQEVPFALSLSSDGAVSPVGELAAGGDRDLESQAGSLVESKARDAAEEVAPPLPMGASVPSIDSMMRKLEEMGLNLRKDQKKTKKRVSFSEQLFTEEAVAGAALLVEGHSSCPQELNPAWSVAGNASDGEPPESPHAEDSERESVTTPGPATCGAPASPADHLLLPSQEESFSEVPMSEASSAKDTPLFRMEGEDALVTQYQSKASDHEGLLSDPLSDLQLVSDFKSPIMADLNLSLPSIPEVASDDERIDQVEDDGDQVEDDGETAKSSTLDIGALSLGLVVPCPERGKGPSGEADRLVLGEGLCDFRLQAPQASVTAPSEQTTEFGIHKPHLGKSSSLDKQLPGPSGGEEEKPMGNGSPSPPPGTSLDNPVPSPSPSEIFPVTHSFPSSAHSDTHHTSTAESQKKATAEGSAGRVENFGKRKPLLQAWVSPSETHPVSAQPGAGTGSAKHRLHPVKPMNAMATKVANCSLGTATIISENLNNEVMMKKYSPSDPAFAYAQLTHDELIQLVLKQKETISKKEFQVRELEDYIDNLLVRVMEETPNILRIPTQVGKKAGKMSpecifications
NP_001002233.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
91.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686E2214/FLJ22524/FLJ22622/MGC78448/NOEL1A/RCP/rab11-FIP1 | |
RAB11FIP1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
80223 | |
RAB11FIP1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGRTRETGTEKNTSSLELEESLPEQPETGRQEEELPRFPCKKQDYSPSSGEAQEVPFALSLSSDGAVSPVGELAAGGDRDLESQAGSLVESKARDAAEEVAPPLPMGASVPSIDSMMRKLEEMGLNLRKDQKKTKKRVSFSEQLFTEEAVAGAALLVEGHSSCPQELNPAWSVAGNASDGEPPESPHAEDSERESVTTPGPATCGAPASPADHLLLPSQEESFSEVPMSEASSAKDTPLFRMEGEDALVTQYQSKASDHEGLLSDPLSDLQLVSDFKSPIMADLNLSLPSIPEVASDDERIDQVEDDGDQVEDDGETAKSSTLDIGALSLGLVVPCPERGKGPSGEADRLVLGEGLCDFRLQAPQASVTAPSEQTTEFGIHKPHLGKSSSLDKQLPGPSGGEEEKPMGNGSPSPPPGTSLDNPVPSPSPSEIFPVTHSFPSSAHSDTHHTSTAESQKKATAEGSAGRVENFGKRKPLLQAWVSPSETHPVSAQPGAGTGSAKHRLHPVKPMNAMATKVANCSLGTATIISENLNNEVMMKKYSPSDPAFAYAQLTHDELIQLVLKQKETISKKEFQVRELEDYIDNLLVRVMEETPNILRIPTQVGKKAGKM | |
RUO | |
RAB11FIP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |