Learn More
ACROBiosystems Human PVRIG Protein, His Tag
Brand:  ACROBiosystems PVG-H52H4-100ug
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16447999
  • 4432.75 NOK / 100µg
Estimated Shipment: 19-01-2023
to see stock.
Description
Description
Human PVRIG (poliovirus receptor related immunoglobulin domain-containing protein), also known as CD112 receptor (CD112R), is an approximately 34 kDa single transmembrane protein in the poliovirus receptor-like protein (PVR) family. The CD112R gene encodes a putative single transmembrane protein, which is composed of a single extracellular IgV domain, one transmembrane domain, and a long intracellular domain. Notably, the intracellular domain of phatases. The extracellular domain sequence of human and mouse CD112R have 65.3% similarity. CD112R may act as a coinhibitory receptor that suppresses T-cell receptor-mediated signals.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 15.6 kDa. The protein migrates as 20-30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Specifications
Specifications
NP_076975.2
15.6 kDa
100 µg
Thr 41 - Asp 171
C7orf15,C7orf15MGC138295,CD112R,MGC104322,MGC138297,MGC2463,PVRIG,CD112 receptor
Unconjugated
PBS, pH7.4
HEK293
Human
Powder
>90% as determined by SDS-PAGE
Human PVRIG, His Tag (PVG-H52H4) is expressed from human 293 cells (HEK293). It contains AA Thr 41 - Asp 171 (Accession # Q6DKI7-1).
-20°C
PVRIG
His Tag
Less than 1.0 EU / µg by the LAL method.
Immune Checkpoint Proteins