missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PTPRO Partial ORF (NP_002839.1, 111 a.a. - 209 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00005800-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16123905

  • 6245.00 NOK / 25µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a receptor-type protein tyrosine phosphatase containing a single intracellular catalytic domain with a characteristic signature motif. The gene product, which has a transmembrane domain, is an integral membrane protein. Several alternatively spliced transcript variants, some of which encode different isoforms of the protein, have been described. These variants exhibit tissue-specific expression. [provided by RefSeq]

Sequence: SITVLTKPLPVTSVSIYDYKPSPETGVLFEIHYPEKYNVFTRVNISYWEGKDFRTMLYKDFFKGKTVFNHWLPGMCYSNITFQLVSEATFNKSTLVEYS
Specifications

Specifications

NP_002839.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SITVLTKPLPVTSVSIYDYKPSPETGVLFEIHYPEKYNVFTRVNISYWEGKDFRTMLYKDFFKGKTVFNHWLPGMCYSNITFQLVSEATFNKSTLVEYS
RUO
PTPRO
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
5800
PTPRO (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GLEPP1/PTP-U2/PTPU2
PTPRO
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers