missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PSRC2 Full-length ORF (ENSP00000307825, 1 a.a. - 358 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00196441-P02.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16189173

  • 6245.00 NOK / 25µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MATADTPAPASSGLSPKEEGELEDGEISDDDNNSQIRSRSSSSSSGGGLLPYPRRRPPHSARGGGSGGGGGSSSSSSSSQQQLRNFSRSRHASERGHLRGPSSYRPKEPFRSHPPSVRMPSSSLSESSPRPSFWERSHLALDRFRFRGRPYRGGSRWSRGRGVGERGGKPGCRPPLGGGAGSGFSSSQSWREPSPPRKSSKSFGRSPSRKQNYSSKNENCVEETFEDLLLKYKQIQLELECINKDEKLALSSKEENVQEDPKTLNFEDQTSTDNVSITKDSSKEVAPEEKTQVKTFQAFELKPLRQKLTLPGDKNRLKKVKDGAKPLSLKSDTTDSSQGIPYRVKEGFTPIPGLKFSA
Specifications

Specifications

ENSP00000307825
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
65.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CCDC131/DKFZp686A0722/KIAA0546/MGC23401/MGC90200/PSRC2
ZFC3H1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
196441
PSRC2 (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue.
MATADTPAPASSGLSPKEEGELEDGEISDDDNNSQIRSRSSSSSSGGGLLPYPRRRPPHSARGGGSGGGGGSSSSSSSSQQQLRNFSRSRHASERGHLRGPSSYRPKEPFRSHPPSVRMPSSSLSESSPRPSFWERSHLALDRFRFRGRPYRGGSRWSRGRGVGERGGKPGCRPPLGGGAGSGFSSSQSWREPSPPRKSSKSFGRSPSRKQNYSSKNENCVEETFEDLLLKYKQIQLELECINKDEKLALSSKEENVQEDPKTLNFEDQTSTDNVSITKDSSKEVAPEEKTQVKTFQAFELKPLRQKLTLPGDKNRLKKVKDGAKPLSLKSDTTDSSQGIPYRVKEGFTPIPGLKFSA
RUO
ZFC3H1
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers