missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PRR4 Partial ORF (NP_009175.1, 17 a.a. - 116 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00011272-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16124016

  • 4115.00 NOK / 10µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Lacrimal proline rich protein is a member of the proline-rich protein family which lacks a conserved repetitive domain. It may have a role in protective functions in the eye. Two alternatively spliced transcript variants that encode different proteins have been described for this gene. [provided by RefSeq]

Sequence: QSTDNDVNYEDFTFTIPDVEDSSQRPDQGPQRPPPEGLLPRPPGDSGNQDDGPQQRPPKPGGHHRHPPPPPFQNQQRPPQRGHRQLSLPRFPSVSLQEAS
Specifications

Specifications

NP_009175.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
QSTDNDVNYEDFTFTIPDVEDSSQRPDQGPQRPPPEGLLPRPPGDSGNQDDGPQQRPPKPGGHHRHPPPPPFQNQQRPPQRGHRQLSLPRFPSVSLQEAS
RUO
PRR4
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
11272
PRR4 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp779L1763/LPRP/PROL4
PRR4
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers