Learn More
Abnova™ Human PRPS1L1 Partial ORF (NP_787082, 146 a.a. - 243 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00221823-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This intronless gene is specifically expressed in the testis, and encodes a protein that is highly homologous to the two subunits of phosphoribosylpyrophosphate synthetase encoded by human X-linked genes, PRPS1 and PRPS2. These enzymes convert pyrimidine, purine or pyridine bases to the corresponding ribonucleoside monophosphates. In vitro transcription/translation and site-directed mutagenesis studies indicate that translation of this mRNA initiates exclusively at a non-AUG (ACG) codon. [provided by RefSeq]
Sequence: YAEPTVLKWIRENIPEWKNCIIVSPDAGGAKRVTSIADQLNVDFALIHKERKKANEVDCIVLVGDVNDRVAILVDDMADTCVTICLAADKLLSAGATRSpecifications
NP_787082 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
YAEPTVLKWIRENIPEWKNCIIVSPDAGGAKRVTSIADQLNVDFALIHKERKKANEVDCIVLVGDVNDRVAILVDDMADTCVTICLAADKLLSAGATR | |
RUO | |
PRPS1L1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
221823 | |
PRPS1L1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PRPS1/PRPS3/PRPSL/PRS-III | |
PRPS1L1 | |
Recombinant | |
wheat germ expression system |