missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PRPF3 Partial ORF (NP_004689, 585 a.a. - 683 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009129-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16189995

  • 4115.00 NOK / 10µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The removal of introns from nuclear pre-mRNAs occurs on complexes called spliceosomes, which are made up of 4 small nuclear ribonucleoprotein (snRNP) particles and an undefined number of transiently associated splicing factors. This gene product is one of several proteins that associate with U4 and U6 snRNPs. Mutations in this gene are associated with retinitis pigmentosa-18. [provided by RefSeq]

Sequence: EGGPKAQKKFKRLMLHRIKWDEQTSNTKGDDDEESDEEAVKKTNKCVLVWEGTAKDRSFGEMKFKQCPTENMAREHFKKHGAEHYWDLALSESVLESTD
Specifications

Specifications

NP_004689
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
EGGPKAQKKFKRLMLHRIKWDEQTSNTKGDDDEESDEEAVKKTNKCVLVWEGTAKDRSFGEMKFKQCPTENMAREHFKKHGAEHYWDLALSESVLESTD
RUO
PRPF3
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
9129
PRPF3 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HPRP3/HPRP3P/PRP3/Prp3p/RP18
PRPF3
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers