missing translation for 'onlineSavingsMsg'
Learn More

Human Pro-Corticoliberin/Pro-CRH Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18703698
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
Product Code. Quantity unitSize
18703698 25 μg 25µg
1 options
This item is not returnable. View return policy

Product Code. 18703698

Brand: R&D Systems AF5805SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Pro-Corticoliberin/Pro-CRH Polyclonal specifically detects Pro-Corticoliberin/Pro-CRH in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pro-Corticoliberin/Pro-CRH
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P06850
Gene Alias CRF, CRH, CRH1, ProCorticoliberin
Gene Symbols CRH
Host Species Sheep
Immunogen E. coli-derived recombinant human Pro-Corticoliberin/Pro-CRH Leu25-Lys196 Accession # P06850
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1392
Test Specificity Detects human Pro-Corticoliberin/Pro-CRH in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.