missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PRIC285 Partial ORF (NP_208384.2, 1973 a.a. - 2078 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00085441-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16141027

  • 6245.00 NOK / 25µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is a nuclear transcriptional co-activator for peroxisome proliferator activated receptor alpha. The encoded protein contains a zinc finger and is a helicase that appears to be part of the peroxisome proliferator activated receptor alpha interacting complex. This gene is a member of the DNA2/NAM7 helicase gene family. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: GVAVSSITKSQGSEWRYVLVSTVRTCAKSDLDQRPTKSWLKKFLGFVVDPNQVNVAVTRAQEGLCLIGDHLLLRCCPLWRSLLDFCEAQQTLVPAGQVRVCRRPTM
Specifications

Specifications

NP_208384.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.4kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
GVAVSSITKSQGSEWRYVLVSTVRTCAKSDLDQRPTKSWLKKFLGFVVDPNQVNVAVTRAQEGLCLIGDHLLLRCCPLWRSLLDFCEAQQTLVPAGQVRVCRRPTM
RUO
PRIC285
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
85441
PRIC285 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ00244/KIAA1769/MGC132634/MGC138228/PDIP-1
PRIC285
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers