missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PPP3R2 Full-length ORF (NP_671709.1, 1 a.a. - 173 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00005535-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16139481

  • 3602.46 NOK / 10µg

Estimated Shipment: 20-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MSTMGNEASYPAEMCSHFDNDEIKRLGRRFKKLDLDKSGSLSVEEFMSLPELRHNPLVRRVIDVFDTDGDGEVDFKEFILGTSQFSVKGDEEQKLRFAFSIYDMDKDGYISNGELFQVLKMMVGNNLTDWQLQQLVDKTIIILDKDGDGKISFEEFSAVVRDLEIHKKLVLIV
Specifications

Specifications

NP_671709.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
46.3kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PPP3RL
PPP3R2
Yes
wheat germ expression system
Liquid
5535
PPP3R2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSTMGNEASYPAEMCSHFDNDEIKRLGRRFKKLDLDKSGSLSVEEFMSLPELRHNPLVRRVIDVFDTDGDGEVDFKEFILGTSQFSVKGDEEQKLRFAFSIYDMDKDGYISNGELFQVLKMMVGNNLTDWQLQQLVDKTIIILDKDGDGKISFEEFSAVVRDLEIHKKLVLIV
RUO
PPP3R2
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates