missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PPP2R5E Full-length ORF (NP_006237.1, 1 a.a. - 467 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00005529-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16199461

  • 6245.00 NOK / 25µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The product of this gene belongs to the phosphatase 2A regulatory subunit B family. Protein phosphatase 2A is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The B regulatory subunit might modulate substrate selectivity and catalytic activity. This gene encodes an epsilon isoform of the regulatory subunit B56 subfamily. [provided by RefSeq]

Sequence: MSSAPTTPPSVDKVDGFSRKSVRKARQKRSQSSSQFRSQGKPIELTPLPLLKDVPSSEQPELFLKKLQQCCVIFDFMDTLSDLKMKEYKRSTLNELVDYITISRGCLTEQTYPEVVRMVSCNIFRTLPPSDSNEFDPEEDEPTLEASWPHLQLVYEFFIRFLESQEFQPSIAKKYIDQKFVLQLLELFDSEDPRERDYLKTVLHRIYGKFLGLRAFIRKQINNIFLRFVYETEHFNGVAELLEILGSIINGFALPLKAEHKQFLVKVLIPLHTVRSLSLFHAQLAYCIVQFLEKDPSLTEPVIRGLMKFWPKTCSQKEVMFLGELEEILDVIEPSQFVKIQEPLFKQIAKCVSSPHFQVAERALYYWNNEYIMSLIEENSNVILPIMFSSLYRISKEHWNPAIVALVYNVLKAFMEMNSTMFDELTATYKSDRQREKKKEKEREELWKKLEDLELKRGLRRDGIIPT
Specifications

Specifications

NP_006237.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
81.1kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PPP2R5E
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
5529
PPP2R5E (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSSAPTTPPSVDKVDGFSRKSVRKARQKRSQSSSQFRSQGKPIELTPLPLLKDVPSSEQPELFLKKLQQCCVIFDFMDTLSDLKMKEYKRSTLNELVDYITISRGCLTEQTYPEVVRMVSCNIFRTLPPSDSNEFDPEEDEPTLEASWPHLQLVYEFFIRFLESQEFQPSIAKKYIDQKFVLQLLELFDSEDPRERDYLKTVLHRIYGKFLGLRAFIRKQINNIFLRFVYETEHFNGVAELLEILGSIINGFALPLKAEHKQFLVKVLIPLHTVRSLSLFHAQLAYCIVQFLEKDPSLTEPVIRGLMKFWPKTCSQKEVMFLGELEEILDVIEPSQFVKIQEPLFKQIAKCVSSPHFQVAERALYYWNNEYIMSLIEENSNVILPIMFSSLYRISKEHWNPAIVALVYNVLKAFMEMNSTMFDELTATYKSDRQREKKKEKEREELWKKLEDLELKRGLRRDGIIPT
RUO
PPP2R5E
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers