Learn More
Abnova™ Human PPP2R5A Full-length ORF (AAH22474, 1 a.a. - 486 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005525-P01.25ug
Additional Details : Weight : 0.00010kg
Description
The product of this gene belongs to the phosphatase 2A regulatory subunit B family. Protein phosphatase 2A is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The B regulatory subunit might modulate substrate selectivity and catalytic activity. This gene encodes an alpha isoform of the regulatory subunit B56 subfamily. [provided by RefSeq]
Sequence: MSSSSPPAGAASAAISASEKVDGFTRKSVRKAQRQKRSQGSSQFRSQGSQAELHPLPQLKDATSNEQQELFCQKLQQCCILFDFMDSVSDLKSKEIKRATLNELVEYVSTNRGVIVESAYSDIVKMISANIFRTLPPSDNPDFDPEEDEPTLEASWPHIQLVYEFFLRFLESPDFRPSIAKRYIDQKFVQQLLELFDSEDPRERDFLKTVLHRIYGKFLGLRAFIRKQINNIFLRFIYETEHFNGVAELLEILGSIINGFALPLKAEHKQFLMKVLIPMHTAKGLALFHAQLAYCVVQFLEKDTTLTEPVIRGLLKFWPKTCSQKEVMFLGEIEEILDVIEPTQFKKIEEPLFKQISKCVSSSHFQVAERALYFWNNEYILSLIEENINKILPIMFASLYKISKEHWNPTIVALVYNVLKTLMEMNGKLFDDLTSSYKAERQREKKKELEREELWKKLEELKLKKALEKQNSAYNMHSILSNTSAESpecifications
AAH22474 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
79.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
B56A/MGC131915/PR61A | |
PPP2R5A | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
5525 | |
PPP2R5A (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSSSSPPAGAASAAISASEKVDGFTRKSVRKAQRQKRSQGSSQFRSQGSQAELHPLPQLKDATSNEQQELFCQKLQQCCILFDFMDSVSDLKSKEIKRATLNELVEYVSTNRGVIVESAYSDIVKMISANIFRTLPPSDNPDFDPEEDEPTLEASWPHIQLVYEFFLRFLESPDFRPSIAKRYIDQKFVQQLLELFDSEDPRERDFLKTVLHRIYGKFLGLRAFIRKQINNIFLRFIYETEHFNGVAELLEILGSIINGFALPLKAEHKQFLMKVLIPMHTAKGLALFHAQLAYCVVQFLEKDTTLTEPVIRGLLKFWPKTCSQKEVMFLGEIEEILDVIEPTQFKKIEEPLFKQISKCVSSSHFQVAERALYFWNNEYILSLIEENINKILPIMFASLYKISKEHWNPTIVALVYNVLKTLMEMNGKLFDDLTSSYKAERQREKKKELEREELWKKLEELKLKKALEKQNSAYNMHSILSNTSAE | |
RUO | |
PPP2R5A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |