Learn More
Abnova™ Human PPP2R2C Partial ORF (NP_065149, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005522-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The product of this gene belongs to the phosphatase 2 regulatory subunit B family. Protein phosphatase 2 is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The B regulatory subunit might modulate substrate selectivity and catalytic activity. This gene encodes a gamma isoform of the regulatory subunit B55 subfamily. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]
Sequence: MGEDTDTRKINHSFLRDHSYVTEADIISTVEFNHTGELLATGDKGGRVVIFQREPESKNAPHSQGEYDVYSTFQSHEPEFDYLKSLEIEEKINKIKWLPQQNAAHSLLSTSpecifications
NP_065149 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGEDTDTRKINHSFLRDHSYVTEADIISTVEFNHTGELLATGDKGGRVVIFQREPESKNAPHSQGEYDVYSTFQSHEPEFDYLKSLEIEEKINKIKWLPQQNAAHSLLST | |
RUO | |
PPP2R2C | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
5522 | |
PPP2R2C (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
B55-GAMMA/IMYPNO/IMYPNO1/MGC33570/PR52/PR55G | |
PPP2R2C | |
Recombinant | |
wheat germ expression system |