missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PPP2R1B Full-length ORF (NP_002707.3, 1 a.a. - 601 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00005519-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16141638

  • 4130.00 NOK / 10µg
Estimated Shipment: 06-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a constant regulatory subunit of protein phosphatase 2. Protein phosphatase 2 is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The constant regulatory subunit A serves as a scaffolding molecule to coordinate the assembly of the catalytic subunit and a variable regulatory B subunit. This gene encodes a beta isoform of the constant regulatory subunit A. Defects in this gene could be the cause of some lung and colon cancers. At least two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MAGASELGTGPGAAGGDGDDSLYPIAVLIDELRNEDVQLRLNSIKKLSTIALALGVERTRSELLPFLTDTIYDEDEVLLALAEQLGNFTGLVGGPDFAHCLLPPLENLATVEETVVRDKAVESLRQISQEHTPVALEAYFVPLVKRLASGDWFTSRTSACGLFSVCYPRASNAVKAEIRQQFRSLCSDDTPMVRRAAASKLGEFAKVLELDSVKSEIVPLFTSLASDEQDSVRLLAVEACVSIAQLLSQDDLETLVMPTLRQAAEDKSWRVRYMVADRFSELQKAMGPKITLNDLIPAFQNLLKDCEAEVRAAAAHKVKELGENLPIEDRETIIMNQILPYIKELVSDTNQHVKSALASVIMGLSTILGKENTIEHLLPLFLAQLKDECPDVRLNIISNLDCVNEVIGIRQLSQSLLPAIVELAEDAKWRVRLAIIEYMPLLAGQLGVEFFDEKLNSLCMAWLVDHVYAIREAATNNLMKLVQKFGTEWAQNTIVPKVLVMANDPNYLHRMTTLFCINALSEACGQEITTKQMLPIVLKMAGDQVANVRFNVAKSLQKIGPILDTNALQGEVKPVLQKLGQDEDMDVKYFAQEAISVLALA
Specifications

Specifications

NP_002707.3
Liquid
5519
PPP2R1B (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAGASELGTGPGAAGGDGDDSLYPIAVLIDELRNEDVQLRLNSIKKLSTIALALGVERTRSELLPFLTDTIYDEDEVLLALAEQLGNFTGLVGGPDFAHCLLPPLENLATVEETVVRDKAVESLRQISQEHTPVALEAYFVPLVKRLASGDWFTSRTSACGLFSVCYPRASNAVKAEIRQQFRSLCSDDTPMVRRAAASKLGEFAKVLELDSVKSEIVPLFTSLASDEQDSVRLLAVEACVSIAQLLSQDDLETLVMPTLRQAAEDKSWRVRYMVADRFSELQKAMGPKITLNDLIPAFQNLLKDCEAEVRAAAAHKVKELGENLPIEDRETIIMNQILPYIKELVSDTNQHVKSALASVIMGLSTILGKENTIEHLLPLFLAQLKDECPDVRLNIISNLDCVNEVIGIRQLSQSLLPAIVELAEDAKWRVRLAIIEYMPLLAGQLGVEFFDEKLNSLCMAWLVDHVYAIREAATNNLMKLVQKFGTEWAQNTIVPKVLVMANDPNYLHRMTTLFCINALSEACGQEITTKQMLPIVLKMAGDQVANVRFNVAKSLQKIGPILDTNALQGEVKPVLQKLGQDEDMDVKYFAQEAISVLALA
RUO
PPP2R1B
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
92.6kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC26454/PR65B
PPP2R1B
Yes
wheat germ expression system
Videos
SDS
Documents
Certificates
Special Offers

Special Offers