missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PPP1R9A Partial ORF (XP_371933, 1090 a.a. - 1188 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00055607-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: PCSTAQTSTRSPCMPFSWFNDSRKGSYSFRNLPAPTSSLQPSPETLISDKKGSKNFTFNDDFSPSSTSSADLSGLGAEPKTPGLSQSLALSSDESLDMISpecifications
XP_371933 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PCSTAQTSTRSPCMPFSWFNDSRKGSYSFRNLPAPTSSLQPSPETLISDKKGSKNFTFNDDFSPSSTSSADLSGLGAEPKTPGLSQSLALSSDESLDMI | |
RUO | |
PPP1R9A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
55607 | |
PPP1R9A (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ20068/KIAA1222/NRB1/NRBI/Neurabin-I | |
PPP1R9A | |
Recombinant | |
wheat germ expression system |