missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PPFIBP2 Partial ORF (NP_003612, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00008495-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16148645

  • 6245.00 NOK / 25µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

PPFIBP2, or liprin-beta-2, belongs to the liprin-beta family. See liprin-beta-1 (MIM 603141) for background on liprins.[supplied by OMIM]

Sequence: MASDASHALEAALEQMDGIIAGTKTGADLSDGTCEPGLASPASYMNPFPVLHLIEDLRLALEMLELPQERAALLSQIPGPTAAYIKEWFEESLSQVNHHS
Specifications

Specifications

NP_003612
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MASDASHALEAALEQMDGIIAGTKTGADLSDGTCEPGLASPASYMNPFPVLHLIEDLRLALEMLELPQERAALLSQIPGPTAAYIKEWFEESLSQVNHHS
RUO
PPFIBP2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
8495
PPFIBP2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Cclp1/DKFZp781K06126/MGC42541
PPFIBP2
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers