Learn More
Abnova™ Human PPAP2C Partial ORF (NP_003703, 114 a.a. - 166 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00008612-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the phosphatidic acid phosphatase (PAP) family. PAPs convert phosphatidic acid to diacylglycerol, and function in de novo synthesis of glycerolipids as well as in receptor-activated signal transduction mediated by phospholipase D. This protein is similar to phosphatidic acid phosphatase type 2A (PPAP2A) and type 2B (PPAP2B). All three proteins contain 6 transmembrane regions, and a consensus N-glycosylation site. This protein has been shown to possess membrane associated PAP activity. Three alternatively spliced transcript variants encoding distinct isoforms have been reported. [provided by RefSeq]
Sequence: DLAKYMIGRLRPNFLAVCDPDWSRVNCSVYVQLEKVCRGNPADVTEARLSFYSSpecifications
NP_003703 | |
Liquid | |
8612 | |
PPAP2C (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
LPP2/PAP-2c/PAP2-g | |
PPAP2C | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
31.57kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DLAKYMIGRLRPNFLAVCDPDWSRVNCSVYVQLEKVCRGNPADVTEARLSFYS | |
RUO | |
PPAP2C | |
Wheat Germ (in vitro) | |
GST |