missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PLP2 Full-length ORF (NP_002659.1, 1 a.a. - 152 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00005355-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16139211

  • 5420.23 NOK / 25µg

Estimated Shipment: 10-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MADSERLSAPGCWAACTNFSRTRKGILLFAEIILCLVILICFSASTPGYSSLSVIEMILAAIFFVVYMCDLHTKIPFINWPWSDFFRTLIAAILYLITSIVVLVERGNHSKIVAGVLGLIATCLFGYDAYVTFPVRQPRHTAAPTDPADGPV
Specifications

Specifications

NP_002659.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
43.1kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
A4/A4-LSB/MGC126187
PLP2
Yes
wheat germ expression system
Liquid
5355
PLP2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MADSERLSAPGCWAACTNFSRTRKGILLFAEIILCLVILICFSASTPGYSSLSVIEMILAAIFFVVYMCDLHTKIPFINWPWSDFFRTLIAAILYLITSIVVLVERGNHSKIVAGVLGLIATCLFGYDAYVTFPVRQPRHTAAPTDPADGPV
RUO
PLP2
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates