missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PLA2G5 Full-length ORF (NP_000920.1, 1 a.a. - 138 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00005322-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16191368

  • 6245.00 NOK / 25µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene is a member of the secretory phospholipase A2 family. It is located in a tightly-linked cluster of secretory phospholipase A2 genes on chromosome 1. The encoded enzyme catalyzes the hydrolysis of membrane phospholipids to generate lysophospholipids and free fatty acids including arachidonic acid. It preferentially hydrolyzes linoleoyl-containing phosphatidylcholine substrates. Secretion of this enzyme is thought to induce inflammatory responses in neighboring cells. Alternatively spliced transcript variants have been found, but their full-length nature has not been determined. [provided by RefSeq]

Sequence: MKGLLPLAWFLACSVPAVQGGLLDLKSMIEKVTGKNALTNYGFYGCYCGWGGRGTPKDGTDWCCWAHDHCYGRLEEKGCNIRTQSYKYRFAWGVVTCEPGPFCHVNLCACDRKLVYCLKRNLRSYNPQYQYFPNILCS
Specifications

Specifications

NP_000920.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
42.1kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp686C2294/GV-PLA2/MGC46205/PLA2-10/hVPLA(2)
PLA2G5
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
5322
PLA2G5 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MKGLLPLAWFLACSVPAVQGGLLDLKSMIEKVTGKNALTNYGFYGCYCGWGGRGTPKDGTDWCCWAHDHCYGRLEEKGCNIRTQSYKYRFAWGVVTCEPGPFCHVNLCACDRKLVYCLKRNLRSYNPQYQYFPNILCS
RUO
PLA2G5
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers