missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PITPNM1 Partial ORF (NP_004901.1, 1147 a.a. - 1244 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_004901.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9600 |
Molecular Weight (g/mol) | 36.52kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16191006
|
Abnova™
H00009600-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16181006
|
Abnova™
H00009600-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
PITPNM1 belongs to a family of membrane-associated phosphatidylinositol transfer domain-containing proteins that share homology with the Drosophila retinal degeneration B (rdgB) protein (Ocaka et al., 2005 [PubMed 15627748]).[supplied by OMIM]
Sequence: RAVRKLQAQCQFLSDGYVAHLGQLEAGSHSHASSGPPRAALGKSSYGVAAPVDFLRKQSQLLRSRGPSQAEREGPGTPPTTLARGKARSISLKLDSEESpecifications
NP_004901.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DRES9/FLJ44997/NIR2/PITPNM/RDGB/RDGB1/RDGBA/RDGBA1/Rd9 | |
PITPNM1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
9600 | |
PITPNM1 (Human) Recombinant Protein (Q01) | |
RAVRKLQAQCQFLSDGYVAHLGQLEAGSHSHASSGPPRAALGKSSYGVAAPVDFLRKQSQLLRSRGPSQAEREGPGTPPTTLARGKARSISLKLDSEE | |
RUO | |
PITPNM1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |