Learn More
Abnova™ Human PIPSL Full-length ORF (AAH68549.1, 1 a.a. - 862 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00266971-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This locus is a transcribed pseudogene with similarity to two adjacent chromosome 1 loci: phosphatidylinositol-4-phosphate 5-kinase, type I, alpha (PIP5K1A, GeneID 8394) and proteasome (prosome, macropain) 26S subunit, non-ATPase, 4 (PSMD4, GeneID 5710). The pseudogene is believed to have occurred from L1 retrotransposition into chromosome 10 of a read-through transcript between the two adjacent loci. Evidence of transcription has been found (PubMed IDs: 16344562 and 17623810), but the latter publication found no evidence that the predicted protein is translated. The NCBI RefSeq Project therefore continues to treat this as a transcribed pseudogene. [provided by RefSeq]
Sequence: MASEVPYASGMPIKKIGHRSVDSSGGTTSSALKGAIQLGITHTVGSLSTKPESDVLMQDFHMVESIFFPSEGSNLTPAHHYNAFRFKTYAPVAFRYFWELFGIRPDDYLYSLCSEPLIELCSSGASGSLFYVSSDDEFIVKTVRHKEAEFLQKLLPGYYINLNQNPRTLLPKFYGLYCVQTGGKNIRIVVMNNLLPRSVKMHIKYDLKGSTYRRRASQKEREKPLPTFKDLDFLQDIPDGLFLDADVHNALCKTLQRDCLVLQSFKIMDYSLLMSIHNIDHAQREPLSSETQYSVDTRRPAPQKALYSTAMESIQGEARRGGTMETDDHMGGIPARNSKGERLLLYIGIIDILQSYRFVKKLEHSWKALIHDGDTVSVHRPGFYAEWFQRFMCNTVFKKIPLKPSPSKKLRSGSSFSQRAGSSGNSCITYKPLVSGEHKAQVTTKAEVEPGVHLGCPDVLPQTPPLEEISEGSPTPDPSFSPLVEETLQMLTTSVDNSEYMGNGDFLPTRLQAQQDAVNTVCHSKTRSNPENNVGLITLDNDCEVLTTLTPDTGRILSKLHTVQPKGKITFCMGIHVAHLALKHRQGNNHKIRIIAFVGNPVEDNEKNLVKLAKCLKKEKVNVDIINFGEEEVNTEKLTAFVNTLNGKDGTGSHLVTVPPGPSLADALISFPILAGEGGAMMGLGASDFEFGVDPSADPELALVLRVFMEEQRQRQEEEARQAAAASAAEAGIATTGTEDSDDALLKMTISQQEFGHTGLPDLSSMTEEEKIVCAMQMSLQGAEFGLAESADIDASSAMDTSEPAKEEDDYDVMQDPEFLQSVLENLPGVDPNNEAIRNAVGSLASQATKDSKKDKKEEDEKSpecifications
AAH68549.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
121.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ33990/MGC87444/PIP5K1L1/PSMD4P2/bA429H9.1 | |
PIPSL | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
266971 | |
PIPSL (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MASEVPYASGMPIKKIGHRSVDSSGGTTSSALKGAIQLGITHTVGSLSTKPESDVLMQDFHMVESIFFPSEGSNLTPAHHYNAFRFKTYAPVAFRYFWELFGIRPDDYLYSLCSEPLIELCSSGASGSLFYVSSDDEFIVKTVRHKEAEFLQKLLPGYYINLNQNPRTLLPKFYGLYCVQTGGKNIRIVVMNNLLPRSVKMHIKYDLKGSTYRRRASQKEREKPLPTFKDLDFLQDIPDGLFLDADVHNALCKTLQRDCLVLQSFKIMDYSLLMSIHNIDHAQREPLSSETQYSVDTRRPAPQKALYSTAMESIQGEARRGGTMETDDHMGGIPARNSKGERLLLYIGIIDILQSYRFVKKLEHSWKALIHDGDTVSVHRPGFYAEWFQRFMCNTVFKKIPLKPSPSKKLRSGSSFSQRAGSSGNSCITYKPLVSGEHKAQVTTKAEVEPGVHLGCPDVLPQTPPLEEISEGSPTPDPSFSPLVEETLQMLTTSVDNSEYMGNGDFLPTRLQAQQDAVNTVCHSKTRSNPENNVGLITLDNDCEVLTTLTPDTGRILSKLHTVQPKGKITFCMGIHVAHLALKHRQGNNHKIRIIAFVGNPVEDNEKNLVKLAKCLKKEKVNVDIINFGEEEVNTEKLTAFVNTLNGKDGTGSHLVTVPPGPSLADALISFPILAGEGGAMMGLGASDFEFGVDPSADPELALVLRVFMEEQRQRQEEEARQAAAASAAEAGIATTGTEDSDDALLKMTISQQEFGHTGLPDLSSMTEEEKIVCAMQMSLQGAEFGLAESADIDASSAMDTSEPAKEEDDYDVMQDPEFLQSVLENLPGVDPNNEAIRNAVGSLASQATKDSKKDKKEEDEK | |
RUO | |
PIPSL | |
Wheat Germ (in vitro) | |
GST | |
Liquid |