Learn More
Abnova™ Human PIK3R6 Full-length ORF (AAI66645.1, 1 a.a. - 754 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00146850-P01.25ug
Additional Details : Weight : 0.00010kg
Description
Phosphoinositide 3-kinase gamma is a lipid kinase that produces the lipid second messenger phosphatidylinositol 3,4,5-trisphosphate. The kinase is composed of a catalytic subunit and one of several regulatory subunits, and is chiefly activated by G protein-coupled receptors. This gene encodes a regulatory subunit, and is distantly related to the phosphoinositide-3-kinase, regulatory subunit 5 gene which is located adjacent to this gene on chromosome 7. The orthologous protein in the mouse binds to both the catalytic subunit and to G(beta/gamma), and mediates activation of the kinase subunit downstream of G protein-coupled receptors. [provided by RefSeq]
Sequence: MESSDVELDLQRSVQAVLRELSTQAPALQSNQGMWRWSLHKKVERDPGKSPVLVRILLRELEKAESQDLRHVIIPLLHTVMYVLTKATGITEELYQRIYAFCTRLLTLPTPYCTVALDCAIRLKTEMAVPGTLYQRMVIAEQNLTNELYPYQERVFLFVDPELVSASVCSALLLEIEAAQAQQTPETCMRHVVSHALQAALGEACHAGALHRKLQASPRRTLEHYFHAVVAALEQMASEASPSREGHVERLEEIYCSLLGPAAGRCGGDLVQERPPSIPLPSPYITFHLWTGEEQLWKELVLFLRPRSQLRLSADLEVLDLQGLRPDRELARVSVLSTDSGIERDLPTGADELPAPGSPEMERAGLQRKGGIKKRAWPLDFLMPGSWDGPPGLHRRTGRPSGDGEMLPGVSRLHTARVLVLGDDRMLGRLAQAYHRLRKRETQKFCLTPRLSLQLYYIPVLAPEKPAASRQPELGELATFLGRVDPWYQSNVNTLCPAIHKLAEMPPSLDTSRTVDPFILDVITYYIRMGTQPIYFQIYTVKIFFSDLSQDPTEDIFLIELKVKIQDSKFPKDGFSPRRRGVAEGPGAELSLCYQKALLSHRPREVTVSLRATGLILKAIPASDTEVSGSSHCPLPAAPVTDHTCLNVNVTEVVKSSNLAGKSFSTVTNTFRTNNIQIQSRDQRLLTLSLDKDDQRTFRDVVRFEVAPCPEPCSGAQKSKAPWLNLHGQQEVEAIKAKPKPLLMPINTFSGIVQSpecifications
AAI66645.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
109.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C17orf38/DKFZp666P158/FLJ34500/HsT41028/p84/p87(PIKAP)/p87PIKAP | |
PIK3R6 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
146850 | |
PIK3R6 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
MESSDVELDLQRSVQAVLRELSTQAPALQSNQGMWRWSLHKKVERDPGKSPVLVRILLRELEKAESQDLRHVIIPLLHTVMYVLTKATGITEELYQRIYAFCTRLLTLPTPYCTVALDCAIRLKTEMAVPGTLYQRMVIAEQNLTNELYPYQERVFLFVDPELVSASVCSALLLEIEAAQAQQTPETCMRHVVSHALQAALGEACHAGALHRKLQASPRRTLEHYFHAVVAALEQMASEASPSREGHVERLEEIYCSLLGPAAGRCGGDLVQERPPSIPLPSPYITFHLWTGEEQLWKELVLFLRPRSQLRLSADLEVLDLQGLRPDRELARVSVLSTDSGIERDLPTGADELPAPGSPEMERAGLQRKGGIKKRAWPLDFLMPGSWDGPPGLHRRTGRPSGDGEMLPGVSRLHTARVLVLGDDRMLGRLAQAYHRLRKRETQKFCLTPRLSLQLYYIPVLAPEKPAASRQPELGELATFLGRVDPWYQSNVNTLCPAIHKLAEMPPSLDTSRTVDPFILDVITYYIRMGTQPIYFQIYTVKIFFSDLSQDPTEDIFLIELKVKIQDSKFPKDGFSPRRRGVAEGPGAELSLCYQKALLSHRPREVTVSLRATGLILKAIPASDTEVSGSSHCPLPAAPVTDHTCLNVNVTEVVKSSNLAGKSFSTVTNTFRTNNIQIQSRDQRLLTLSLDKDDQRTFRDVVRFEVAPCPEPCSGAQKSKAPWLNLHGQQEVEAIKAKPKPLLMPINTFSGIVQ | |
RUO | |
PIK3R6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |