missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PIK3R6 Full-length ORF (AAI66645.1, 1 a.a. - 754 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00146850-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16188433

  • 4115.00 NOK / 10µg
Estimated Shipment: 26-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Phosphoinositide 3-kinase gamma is a lipid kinase that produces the lipid second messenger phosphatidylinositol 3,4,5-trisphosphate. The kinase is composed of a catalytic subunit and one of several regulatory subunits, and is chiefly activated by G protein-coupled receptors. This gene encodes a regulatory subunit, and is distantly related to the phosphoinositide-3-kinase, regulatory subunit 5 gene which is located adjacent to this gene on chromosome 7. The orthologous protein in the mouse binds to both the catalytic subunit and to G(beta/gamma), and mediates activation of the kinase subunit downstream of G protein-coupled receptors. [provided by RefSeq]

Sequence: MESSDVELDLQRSVQAVLRELSTQAPALQSNQGMWRWSLHKKVERDPGKSPVLVRILLRELEKAESQDLRHVIIPLLHTVMYVLTKATGITEELYQRIYAFCTRLLTLPTPYCTVALDCAIRLKTEMAVPGTLYQRMVIAEQNLTNELYPYQERVFLFVDPELVSASVCSALLLEIEAAQAQQTPETCMRHVVSHALQAALGEACHAGALHRKLQASPRRTLEHYFHAVVAALEQMASEASPSREGHVERLEEIYCSLLGPAAGRCGGDLVQERPPSIPLPSPYITFHLWTGEEQLWKELVLFLRPRSQLRLSADLEVLDLQGLRPDRELARVSVLSTDSGIERDLPTGADELPAPGSPEMERAGLQRKGGIKKRAWPLDFLMPGSWDGPPGLHRRTGRPSGDGEMLPGVSRLHTARVLVLGDDRMLGRLAQAYHRLRKRETQKFCLTPRLSLQLYYIPVLAPEKPAASRQPELGELATFLGRVDPWYQSNVNTLCPAIHKLAEMPPSLDTSRTVDPFILDVITYYIRMGTQPIYFQIYTVKIFFSDLSQDPTEDIFLIELKVKIQDSKFPKDGFSPRRRGVAEGPGAELSLCYQKALLSHRPREVTVSLRATGLILKAIPASDTEVSGSSHCPLPAAPVTDHTCLNVNVTEVVKSSNLAGKSFSTVTNTFRTNNIQIQSRDQRLLTLSLDKDDQRTFRDVVRFEVAPCPEPCSGAQKSKAPWLNLHGQQEVEAIKAKPKPLLMPINTFSGIVQ
Specifications

Specifications

AAI66645.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
109.4kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C17orf38/DKFZp666P158/FLJ34500/HsT41028/p84/p87(PIKAP)/p87PIKAP
PIK3R6
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
146850
PIK3R6 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue
MESSDVELDLQRSVQAVLRELSTQAPALQSNQGMWRWSLHKKVERDPGKSPVLVRILLRELEKAESQDLRHVIIPLLHTVMYVLTKATGITEELYQRIYAFCTRLLTLPTPYCTVALDCAIRLKTEMAVPGTLYQRMVIAEQNLTNELYPYQERVFLFVDPELVSASVCSALLLEIEAAQAQQTPETCMRHVVSHALQAALGEACHAGALHRKLQASPRRTLEHYFHAVVAALEQMASEASPSREGHVERLEEIYCSLLGPAAGRCGGDLVQERPPSIPLPSPYITFHLWTGEEQLWKELVLFLRPRSQLRLSADLEVLDLQGLRPDRELARVSVLSTDSGIERDLPTGADELPAPGSPEMERAGLQRKGGIKKRAWPLDFLMPGSWDGPPGLHRRTGRPSGDGEMLPGVSRLHTARVLVLGDDRMLGRLAQAYHRLRKRETQKFCLTPRLSLQLYYIPVLAPEKPAASRQPELGELATFLGRVDPWYQSNVNTLCPAIHKLAEMPPSLDTSRTVDPFILDVITYYIRMGTQPIYFQIYTVKIFFSDLSQDPTEDIFLIELKVKIQDSKFPKDGFSPRRRGVAEGPGAELSLCYQKALLSHRPREVTVSLRATGLILKAIPASDTEVSGSSHCPLPAAPVTDHTCLNVNVTEVVKSSNLAGKSFSTVTNTFRTNNIQIQSRDQRLLTLSLDKDDQRTFRDVVRFEVAPCPEPCSGAQKSKAPWLNLHGQQEVEAIKAKPKPLLMPINTFSGIVQ
RUO
PIK3R6
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers