Learn More
Abnova™ Human PIAS2 Partial ORF (NP_004662, 385 a.a. - 473 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009063-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein involved in the regulation of transcription factors involved in MAP kinase signaling. The symbol MIZ1 has also been associated with ZBTB17 which is a different gene located on chromosome 1. Two alternatively spliced transcripts encoding different isoforms have been described. [provided by RefSeq]
Sequence: CPVCDKKAAYESLILDGLFMEILNDCSDVDEIKFQEDGSWCPMRPKKEAMKVSSQPCTKIESSSVLSKPCSVTVASEASKKKVDVIDLTSpecifications
NP_004662 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.53kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
CPVCDKKAAYESLILDGLFMEILNDCSDVDEIKFQEDGSWCPMRPKKEAMKVSSQPCTKIESSSVLSKPCSVTVASEASKKKVDVIDLT | |
RUO | |
PIAS2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9063 | |
PIAS2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC102682/MIZ1/PIASX/PIASX-ALPHA/PIASX-BETA/SIZ2/ZMIZ4/miz | |
PIAS2 | |
Recombinant | |
wheat germ expression system |