missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PI15 Full-length ORF (NP_056970.1, 1 a.a. - 258 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00051050-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16189552

  • 4115.00 NOK / 10µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a trypsin inhibitor. The protein shares similarity to insect venom allergens, mammalian testis-specific proteins and plant pathogenesis-related proteins. It is frequently expressed in human neuroblastoma and glioblastoma cell lines, and thus may play a role in the central nervous system. [provided by RefSeq]

Sequence: MIAISAVSSALLFSLLCEASTVVLLNSTDSSPPTNNFTDIEAALKAQLDSADIPKARRKRYISQNDMIAILDYHNQVRGKVFPPAANMEYMVWDENLAKSAEAWAATCIWDHGPSYLLRFLGQNLSVRTGRYRSILQLVKPWYDEVKDYAFPYPQDCNPRCPMRCFGPMCTHYTQMVWATSNRIGCAIHTCQNMNVWGSVWRRAVYLVCNYAPKGNWIGEAPYKVGVPCSSCPPSYGGSCTDNLCFPGVTSNYLYWFK
Specifications

Specifications

NP_056970.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
55.5kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CRISP8/DKFZp686F0366/P24TI/P25TI
PI15
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
51050
PI15 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MIAISAVSSALLFSLLCEASTVVLLNSTDSSPPTNNFTDIEAALKAQLDSADIPKARRKRYISQNDMIAILDYHNQVRGKVFPPAANMEYMVWDENLAKSAEAWAATCIWDHGPSYLLRFLGQNLSVRTGRYRSILQLVKPWYDEVKDYAFPYPQDCNPRCPMRCFGPMCTHYTQMVWATSNRIGCAIHTCQNMNVWGSVWRRAVYLVCNYAPKGNWIGEAPYKVGVPCSSCPPSYGGSCTDNLCFPGVTSNYLYWFK
RUO
PI15
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers