Learn More
Abnova™ Human PHF7 Full-length ORF (NP_057567.3, 1 a.a. - 381 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00051533-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Spermatogenesis is a complex process regulated by extracellular and intracellular factors as well as cellular interactions among interstitial cells of the testis, Sertoli cells, and germ cells. In the testis, this gene is expressed in Sertoli cells but not germ cells. However, this gene is not expressed in a patient who exhibited spermatogenic arrest at the spermatocyte stage. Spermatogenic arrest is an interruption of germ cell differentiation that may result in oligospermia or azoospermia. The proteins encoded by this gene contain plant homeodomain (PHD) finger domains, also known as leukemia associated protein (LAP) domains, believed to be involved in transcriptional regulation. Thus this protein, which localizes to the nucleus of transfected cells, has been implicated in the transcriptional regulation of spermatogenesis. Two protein isoforms are encoded by transcript variants of this gene. [provided by RefSeq]
Sequence: MKTVKEKKECQRLRKSAKTRRVTQRKPSSGPVCWLCLREPGDPEKLGEFLQKDNISVHYFCLILSSKLPQRGQSNRGFHGFLPEDIKKEAARASRKICFVCKKKGAAINCQKDQCLRNFHLPCGQERGCLSQFFGEYKSFCDKHRPTQNIQHGHVGEESCILCCEDLSQQSVENIQSPCCSQAIYHRKCIQKYAHTSAKHFFKCPQCNNRKEFPQEMLRMGIHIPDRDAAWELEPGAFSDLYQRYQHCDAPICLYEQGRDSFEDEGRWCLILCATCGSHGTHRDCSSLRSNSKKWECEECSPAAATDYIPENSGDIPCCSSTFHPEEHFCRDNTLEENPGLSWTDWPEPSLLEKPESSRGRRSYSWRSKGVRITNSCKKSKSpecifications
NP_057567.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
70.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp434L1850/HSPC045/HSPC226/MGC26088/NYD-SP6 | |
PHF7 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
51533 | |
PHF7 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKTVKEKKECQRLRKSAKTRRVTQRKPSSGPVCWLCLREPGDPEKLGEFLQKDNISVHYFCLILSSKLPQRGQSNRGFHGFLPEDIKKEAARASRKICFVCKKKGAAINCQKDQCLRNFHLPCGQERGCLSQFFGEYKSFCDKHRPTQNIQHGHVGEESCILCCEDLSQQSVENIQSPCCSQAIYHRKCIQKYAHTSAKHFFKCPQCNNRKEFPQEMLRMGIHIPDRDAAWELEPGAFSDLYQRYQHCDAPICLYEQGRDSFEDEGRWCLILCATCGSHGTHRDCSSLRSNSKKWECEECSPAAATDYIPENSGDIPCCSSTFHPEEHFCRDNTLEENPGLSWTDWPEPSLLEKPESSRGRRSYSWRSKGVRITNSCKKSK | |
RUO | |
PHF7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |