missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PGA5 Partial ORF (NP_055039, 203 a.a. - 306 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005222-Q01S
Additional Details : Weight : 0.00010kg
Description
Sequence: WNQGLVSQDLFSVYLSADDKSGSVVIFGGIDSSYYTGSLNWVPVTVEGYWQITVDSITMNGETIACAEGCQAIVDTGTSLLTGPTSPIANIQSDIGASENSDGDSpecifications
NP_055039 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.18kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
WNQGLVSQDLFSVYLSADDKSGSVVIFGGIDSSYYTGSLNWVPVTVEGYWQITVDSITMNGETIACAEGCQAIVDTGTSLLTGPTSPIANIQSDIGASENSDGD | |
RUO | |
PGA5 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
5222 | |
PGA5 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PGA5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |