Learn More
Human PDGF-AA Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB221-500
 View more versions of this product
Product Code. 18721199
  • 18260.00 NOK / 500µg
Description
PDGF-AA Monoclonal specifically detects PDGF-AA in Human samples. It is validated for Neutralization.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PDGF-AA
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
PDGF1, PDGF-A, PDGFAA
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
5154
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG1
Neutralization
114506
Neutralization 0.2-1.0 ug/mL
P04085
PDGFA
E. coli-derived recombinant human PDGF-AA Ser87-Thr211 Accession # P04085
500 μg
Primary
Detects human PDGF-AA in direct ELISAs. In direct ELISAs, approximately 20% cross-reactivity with recombinant human (rh) PDGF-AB is observed and no cross-reactivity with rhPDGF-BB, rhPlGF, rhVEGF206, rhVEGF B 186 rhVEGF C, rhVEGF D, or rhCTGF is observed.
Human
Supernatant
Certificates
Promotions
Promotions
For Research Use Only