Learn More
Abnova™ Human PDE8A Full-length ORF (NP_002596.1, 1 a.a. - 829 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005151-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Phosphodiesterases (PDEs) regulate the intracellular levels of cAMP and cGMP. These cyclic nucleotides play an important role as second messengers in multiple physiologic processes, including regulation of vascular resistance, cardiac output, visceral motility, immune response, inflammation, neuroplasticity, vision, and reproduction. PDEs comprise a large superfamily of enzymes divided into 10 families. Different PDEs can be distinguished by their structure, tissue expression, localization, substrate specificity, regulation, and sensitivity to PDE inhibitors. Diversity in structure and specificity of function make PDEs promising targets for the pharmacotherapy of diseases modulated by cyclic nucleotide signaling (Hetman et al., MIM 2000). See MIM 171885.[supplied by OMIM]
Sequence: MGCAPSIHISERLVAEDAPSPAAPPLSSGGPRLPQGQKTAALPRTRGAGLLESELRDGSGKKVAVADVQFGPMRFHQDQLQVLLVFTKEDNQCNGFCRACEKAGFKCTVTKEAQAVLACFLDKHHDIIIIDHRNPRQLDAEALCRSIRSSKLSENTVIVGVVRRVDREELSVMPFISAGFTRRYVENPNIMACYNELLQLEFGEVRSQLKLRACNSVFTALENSEDAIEITSEDRFIQYANPAFETTMGYQSGELIGKELGEVPINEKKADLLDTINSCIRIGKEWQGIYYAKKKNGDNIQQNVKIIPVIGQGGKIRHYVSIIRVCNGNNKAEKISECVQSDTHTDNQTGKHKDRRKGSLDVKAVASRATEVSSQRRHSSMARIHSMTIEAPITKVINIINAAQESSPMPVTEALDRVLEILRTTELYSPQFGAKDDDPHANDLVGGLMSDGLRRLSGNEYVLSTKNTQMVSSNIITPISLDDVPPRIARAMENEEYWDFDIFELEAATHNRPLIYLGLKMFARFGICEFLHCSESTLRSWLQIIEANYHSSNPYHNSTHSADVLHATAYFLSKERIKETLDPIDEVAALIAATIHDVDHPGRTNSFLCNAGSELAILYNDTAVLESHHAALAFQLTTGDDKCNIFKNMERNDYRTLRQGIIDMVLATEMTKHFEHVNKFVNSINKPLATLEENGETDKNQEVINTMLRTPENRTLIKRMLIKCADVSNPCRPLQYCIEWAARISEEYFSQTDEEKQQGLPVVMPVFDRNTCSIPKSQISFIDYFITDMFDAWDAFVDLPDLMQHLDNNFKYWKGLDEMKLRNLRPPPESpecifications
NP_002596.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
119.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ16150/HsT19550 | |
PDE8A | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
5151 | |
PDE8A (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGCAPSIHISERLVAEDAPSPAAPPLSSGGPRLPQGQKTAALPRTRGAGLLESELRDGSGKKVAVADVQFGPMRFHQDQLQVLLVFTKEDNQCNGFCRACEKAGFKCTVTKEAQAVLACFLDKHHDIIIIDHRNPRQLDAEALCRSIRSSKLSENTVIVGVVRRVDREELSVMPFISAGFTRRYVENPNIMACYNELLQLEFGEVRSQLKLRACNSVFTALENSEDAIEITSEDRFIQYANPAFETTMGYQSGELIGKELGEVPINEKKADLLDTINSCIRIGKEWQGIYYAKKKNGDNIQQNVKIIPVIGQGGKIRHYVSIIRVCNGNNKAEKISECVQSDTHTDNQTGKHKDRRKGSLDVKAVASRATEVSSQRRHSSMARIHSMTIEAPITKVINIINAAQESSPMPVTEALDRVLEILRTTELYSPQFGAKDDDPHANDLVGGLMSDGLRRLSGNEYVLSTKNTQMVSSNIITPISLDDVPPRIARAMENEEYWDFDIFELEAATHNRPLIYLGLKMFARFGICEFLHCSESTLRSWLQIIEANYHSSNPYHNSTHSADVLHATAYFLSKERIKETLDPIDEVAALIAATIHDVDHPGRTNSFLCNAGSELAILYNDTAVLESHHAALAFQLTTGDDKCNIFKNMERNDYRTLRQGIIDMVLATEMTKHFEHVNKFVNSINKPLATLEENGETDKNQEVINTMLRTPENRTLIKRMLIKCADVSNPCRPLQYCIEWAARISEEYFSQTDEEKQQGLPVVMPVFDRNTCSIPKSQISFIDYFITDMFDAWDAFVDLPDLMQHLDNNFKYWKGLDEMKLRNLRPPPE | |
RUO | |
PDE8A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |