missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PDE6C Partial ORF (NP_006195, 3 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005146-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: EINQVAVEKYLEENPQFAKEYFDRKLRVEVLGEIFKNSQVPVQSSMSFSELTQVEESALCLELLWTVQEEGGTPEQGVHRALQRLAHLLQADRCSMFLSpecifications
NP_006195 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
EINQVAVEKYLEENPQFAKEYFDRKLRVEVLGEIFKNSQVPVQSSMSFSELTQVEESALCLELLWTVQEEGGTPEQGVHRALQRLAHLLQADRCSMFL | |
RUO | |
PDE6C | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
5146 | |
PDE6C (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PDEA2 | |
PDE6C | |
Recombinant | |
wheat germ expression system |