missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PCDH8 Partial ORF (NP_002581, 32 a.a. - 120 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_002581 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 5100 |
Molecular Weight (g/mol) | 35.53kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16198761
|
Abnova™
H00005100-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16188761
|
Abnova™
H00005100-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene belongs to the protocadherin gene family, a subfamily of the cadherin superfamily. The gene encodes an integral membrane protein that is thought to function in cell adhesion in a CNS-specific manner. Unlike classical cadherins, which are generally encoded by 15-17 exons, this gene includes only 3 exons. Notable is the large first exon encoding the extracellular region, including 6 cadherin domains and a transmembrane region. Alternative splicing yields isoforms with unique cytoplasmic tails. [provided by RefSeq]
Sequence: VRYSTFEEDAPGTVIGTLAEDLHMKVSGDTSFRLMKQFNSSLLRVREGDGQLTVGDAGLDRERLCGQAPQCVLAFDVVSFSQEQFRLVHSpecifications
NP_002581 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.53kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARCADLIN/PAPC | |
PCDH8 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
5100 | |
PCDH8 (Human) Recombinant Protein (Q01) | |
VRYSTFEEDAPGTVIGTLAEDLHMKVSGDTSFRLMKQFNSSLLRVREGDGQLTVGDAGLDRERLCGQAPQCVLAFDVVSFSQEQFRLVH | |
RUO | |
PCDH8 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |