Learn More
Abnova™ Human PCDH1 Full-length ORF (AAH51287.1, 1 a.a. - 345 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005097-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene belongs to the protocadherin subfamily within the cadherin superfamily. The encoded protein is a membrane protein found at cell-cell boundaries. It is involved in neural cell adhesion, suggesting a possible role in neuronal development. The protein includes an extracelllular region, containing 7 cadherin-like domains, a transmembrane region and a C-terminal cytoplasmic region. Cells expressing the protein showed cell aggregation activity. Alternative splicing occurs in this gene. [provided by RefSeq]
Sequence: MDSGAGGRRCPEAALLILGPPRMEHLRHSPGPGGQRLLLPSMLLALLLLLAPSPGHATRVVYKVPEEQPPNTLIGSLAADYGFPDVGHLYKLEVGAPYLRVDGKTGDIFTTETSIDREGLRECQNQLPGDPCILEFEVSITDLVQNGSPRLLEGQIEVQDINDNTPNFASPVITLAIPENTNIGSLFPIPLASDRDAGPNGVASYELQAGPEAQELFGLQVAEDQEEKQPQLIVMGNLDRERWDSYDLTIKVQDGGSPPRASSALLRVTVLDTNDNAPKFERPSYEAELSENSPIGHSVIQVKANDSDQGANAEIEYTFHQAPEVVRRLLRLDLSLFRARWTVRTSpecifications
AAH51287.1 | |
Liquid | |
5097 | |
PCDH1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MDSGAGGRRCPEAALLILGPPRMEHLRHSPGPGGQRLLLPSMLLALLLLLAPSPGHATRVVYKVPEEQPPNTLIGSLAADYGFPDVGHLYKLEVGAPYLRVDGKTGDIFTTETSIDREGLRECQNQLPGDPCILEFEVSITDLVQNGSPRLLEGQIEVQDINDNTPNFASPVITLAIPENTNIGSLFPIPLASDRDAGPNGVASYELQAGPEAQELFGLQVAEDQEEKQPQLIVMGNLDRERWDSYDLTIKVQDGGSPPRASSALLRVTVLDTNDNAPKFERPSYEAELSENSPIGHSVIQVKANDSDQGANAEIEYTFHQAPEVVRRLLRLDLSLFRARWTVRT | |
RUO | |
PCDH1 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
64.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ53887/MGC45991/PC42/PCDH42 | |
PCDH1 | |
Yes | |
wheat germ expression system |