missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PBK Partial ORF (NP_060962.2, 122 a.a. - 229 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00055872-Q02.25ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16035575

  • 6245.00 NOK / 25µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This genes encodes a serine/threonine kinase related to the dual specific mitogen-activated protein kinase kinase (MAPKK) family. Evidence suggests that mitotic phosphorylation is required for its catalytic activity. This mitotic kinase may be involved in the activation of lymphoid cells and support testicular functions, with a suggested role in the process of spermatogenesis. [provided by RefSeq]

Sequence: SLNDLIEERYKASQDPFPAAIILKVALNMARGLKYLHQEKKLLHGDIKSSNVVIKGDFETIKICDVGVSLPLDENMTVTDPEACYIGTEPWKPKEAVEENGVITDKAD
Specifications

Specifications

NP_060962.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.62kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SLNDLIEERYKASQDPFPAAIILKVALNMARGLKYLHQEKKLLHGDIKSSNVVIKGDFETIKICDVGVSLPLDENMTVTDPEACYIGTEPWKPKEAVEENGVITDKAD
RUO
PBK
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
55872
PBK (Human) Recombinant Protein (Q02)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ14385/Nori-3/SPK/TOPK
PBK
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers