Learn More
Abnova™ Human PAX4 Partial ORF (NP_006184, 121 a.a. - 217 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005078-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the paired box (PAX) family of transcription factors. Members of this gene family typically contain a paired box domain, an octapeptide, and a paired-type homeodomain. These genes play critical roles during fetal development and cancer growth. The paired box 4 gene is involved in pancreatic islet development and mouse studies have demonstrated a role for this gene in differentiation of insulin-producing beta cells. [provided by RefSeq]
Sequence: RALQEDQGLPCTRLRSPAVLAPAVLTPHSGSETPRGTHPGTGHRNRTIFSPSQAEALEKEFQRGQYPDSVARGKLATATSLPEDTVRVWFSNRRAKWSpecifications
NP_006184 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.41kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RALQEDQGLPCTRLRSPAVLAPAVLTPHSGSETPRGTHPGTGHRNRTIFSPSQAEALEKEFQRGQYPDSVARGKLATATSLPEDTVRVWFSNRRAKW | |
RUO | |
PAX4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
5078 | |
PAX4 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KPD/MGC129960/MODY9 | |
PAX4 | |
Recombinant | |
wheat germ expression system |