Learn More
Abnova™ Human PAPPA Partial ORF (NP_002572, 85 a.a. - 194 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005069-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a secreted metalloproteinase which cleaves insulin-like growth factor binding proteins (IGFBPs). It is thought to be involved in local proliferative processes such as wound healing and bone remodeling. Low plasma level of this protein has been suggested as a biochemical marker for pregnancies with aneuploid fetuses. [provided by RefSeq]
Sequence: GATEEPSPPSRALYFSGRGEQLRLRADLELPRDAFTLQVWLRAEGGQRSPAVITGLYDKCSYISRDRGWVVGIHTISDQDNKDPRYFFSLKTDRARQVTTINAHRSYLPGSpecifications
NP_002572 | |
Liquid | |
5069 | |
PAPPA (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ASBABP2/DIPLA1/IGFBP-4ase/PAPA/PAPP-A/PAPPA1 | |
PAPPA | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GATEEPSPPSRALYFSGRGEQLRLRADLELPRDAFTLQVWLRAEGGQRSPAVITGLYDKCSYISRDRGWVVGIHTISDQDNKDPRYFFSLKTDRARQVTTINAHRSYLPG | |
RUO | |
PAPPA | |
Wheat Germ (in vitro) | |
GST |