missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human OVOL2 Full-length ORF (NP_067043.2, 1 a.a. - 275 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00058495-P02.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MPKVFLVKRRSLGVSVRSWDELPDEKRADTYIPVGLGRLLHDPPEDCRSDGGSSSGSGSSSAGEPGGAESSSSPHAPESETPEPGDAEGPDGHLATKQRPVARSKIKFTTGTCSDSVVHSCDLCGKGFRLQRMLNRHLKCHNQVKRHLCTFCGKGFNDTFDLKRHVRTHTGIRPYKCNVCNKAFTQRCSLESHLKKIHGVQQQYAYKQRRDKLYVCEDCGYTGPTQEDLYLHVNSAHPGSSFLKKTSKKLAALLQGKLTSAHQENTSLSEEEERKSpecifications
NP_067043.2 | |
Liquid | |
58495 | |
OVOL2 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MPKVFLVKRRSLGVSVRSWDELPDEKRADTYIPVGLGRLLHDPPEDCRSDGGSSSGSGSSSAGEPGGAESSSSPHAPESETPEPGDAEGPDGHLATKQRPVARSKIKFTTGTCSDSVVHSCDLCGKGFRLQRMLNRHLKCHNQVKRHLCTFCGKGFNDTFDLKRHVRTHTGIRPYKCNVCNKAFTQRCSLESHLKKIHGVQQQYAYKQRRDKLYVCEDCGYTGPTQEDLYLHVNSAHPGSSFLKKTSKKLAALLQGKLTSAHQENTSLSEEEERK | |
RUO | |
OVOL2 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
56.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
EUROIMAGE566589/ZNF339 | |
OVOL2 | |
Yes | |
wheat germ expression system |