Learn More
Abnova™ Human OVOL1 Full-length ORF (AAH59408, 1 a.a. - 267 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005017-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein highly similar to Drosophila and mouse proteins. In Drosophila the ovo protein plays a critical role in Drosophila oogenesis and cuticle formation. In mice the ovo like protein is involved in hair formation and spermatogenesis. The function of the human gene product has not been determined. [provided by RefSeq]
Sequence: MPRAFLVKKPCVSTCKRNWSELPDEGRGEIYVPVSLGFCPPQPYREPEPSVAEPPSCPLALNMSLRDSSYSMAPGPCVVAQLPSEDMGHLTDPQSRDHGFLRAKMKVTLGDSPSGDLFTCRVCQRAFTYQRMLNRHMKCHNDVKRHLCTYCGKGFNDTFDLKRHVRTHTGVRPYKCSLCDKAFTQRCSLESHLKKIHGVQQKYAYKERRAKLYVCEECGCTSESQEGHVLHLKEHHPDSPLLRKTSKKVAVALQNTVTSLLQGSPHLSpecifications
AAH59408 | |
Liquid | |
5017 | |
OVOL1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MPRAFLVKKPCVSTCKRNWSELPDEGRGEIYVPVSLGFCPPQPYREPEPSVAEPPSCPLALNMSLRDSSYSMAPGPCVVAQLPSEDMGHLTDPQSRDHGFLRAKMKVTLGDSPSGDLFTCRVCQRAFTYQRMLNRHMKCHNDVKRHLCTYCGKGFNDTFDLKRHVRTHTGVRPYKCSLCDKAFTQRCSLESHLKKIHGVQQKYAYKERRAKLYVCEECGCTSESQEGHVLHLKEHHPDSPLLRKTSKKVAVALQNTVTSLLQGSPHL | |
RUO | |
OVOL1 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
55.11kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HOVO1 | |
OVOL1 | |
Yes | |
wheat germ expression system |