missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human OTUD5 Full-length ORF (AAH98440.1, 1 a.a. - 542 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00055593-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16181599

  • 4115.00 NOK / 10µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the OTU (ovarian tumor) domain-containing cysteine protease superfamily. The OTU domain confers deubiquitinase activity and the encoded protein has been shown to suppress the type I interferon-dependent innate immune response by cleaving the polyubiquitin chain from an essential type I interferon adaptor protein. Cleavage results in disassociation of the adaptor protein from a downstream signaling complex and disruption of the type I interferon signaling cascade. Alternatively spliced transcript variants encoding different isoforms have been described. [provided by RefSeq]

Sequence: MTILPKKKPPPPDADPGGTGVGGGDRDRDSGVVGARPRASPPPQGPLPGPPGALHRWALAVPPGAVAGPRPQQASPPPCGGPGGPGGGPGDALGAAAAGVGAAGVVVGVGGAVGVGGCCSGPGHSKRRRQAPGVGAVGGGSPEREEVGAGYNSEDEYEAAAARIEAMDPATVEQQEHWFEKALRDKKGFIIKQMKEDGACLFRAVADQVYGDQDMHEVVRKHCMDYLMKNADYFSNYVTEDFTTYINRKRKNNCHGNHIEMQAMAEMYNRPVEVYQYSTEPINTFHGIHQNEDEPIRVSYHRNIHYNSVVNPNKATIGVGLGLPSFKPGFAEQSLMKNAIKTSEESWIEQQMLEDKKRATDWEATNEAIEEQVARESYLQWLRDQEKQARQVRGPSQPRKASATCSSATAAASSGLEEWTSRSPRQRSSASSPEHPELHAELGMKPPSPGTVLALAKPPSPCAPGTSSQFSAGADRATSPLVSLYPALECRALIQQMSPSAFGLNDWDDDEILASVLAVSQQEYLDSMKKNKVHRDPPPDKS
Specifications

Specifications

AAH98440.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
84.4kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp761A052/DUBA/MGC104871
OTUD5
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
55593
OTUD5 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MTILPKKKPPPPDADPGGTGVGGGDRDRDSGVVGARPRASPPPQGPLPGPPGALHRWALAVPPGAVAGPRPQQASPPPCGGPGGPGGGPGDALGAAAAGVGAAGVVVGVGGAVGVGGCCSGPGHSKRRRQAPGVGAVGGGSPEREEVGAGYNSEDEYEAAAARIEAMDPATVEQQEHWFEKALRDKKGFIIKQMKEDGACLFRAVADQVYGDQDMHEVVRKHCMDYLMKNADYFSNYVTEDFTTYINRKRKNNCHGNHIEMQAMAEMYNRPVEVYQYSTEPINTFHGIHQNEDEPIRVSYHRNIHYNSVVNPNKATIGVGLGLPSFKPGFAEQSLMKNAIKTSEESWIEQQMLEDKKRATDWEATNEAIEEQVARESYLQWLRDQEKQARQVRGPSQPRKASATCSSATAAASSGLEEWTSRSPRQRSSASSPEHPELHAELGMKPPSPGTVLALAKPPSPCAPGTSSQFSAGADRATSPLVSLYPALECRALIQQMSPSAFGLNDWDDDEILASVLAVSQQEYLDSMKKNKVHRDPPPDKS
RUO
OTUD5
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers