missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human OSBPL11 Partial ORF (NP_073613.2, 649 a.a. - 746 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00114885-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16111447

  • 4115.00 NOK / 10µg
Estimated Shipment: 02-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Like most members, the encoded protein contains an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain. [provided by RefSeq]

Sequence: KYVDLTKLAVTKKRVRPLEKQDPFESRRLWKNVTDSLRESEIDKATEHKHTLEERQRTEERHRTETGTPWKTKYFIKEGDGWVYHKPLWKIIPTTQPA
Specifications

Specifications

NP_073613.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
KYVDLTKLAVTKKRVRPLEKQDPFESRRLWKNVTDSLRESEIDKATEHKHTLEERQRTEERHRTETGTPWKTKYFIKEGDGWVYHKPLWKIIPTTQPA
RUO
OSBPL11
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
114885
OSBPL11 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ13012/FLJ13164/ORP-11/ORP11/OSBP12/TCCCIA00292
OSBPL11
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers