missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human OR2C3 Partial ORF (NP_932340.2, 220 a.a. - 319 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00081472-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16140387

  • 5540.00 NOK / 25µg
Estimated Shipment: 23-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms. [provided by RefSeq]

Sequence: HIARAVLKIRSAEGRRKAFNTCSSHVAVVSLFYGSIIFMYLQPAKSTSHEQGKFIALFYTVVTPALNPLIYTLRNTEVKSALRHMVLENCCGSAGKLAQI
Specifications

Specifications

NP_932340.2
Liquid
81472
OR2C3 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
OR2C4/OR2C5P/OST742
OR2C3
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue
HIARAVLKIRSAEGRRKAFNTCSSHVAVVSLFYGSIIFMYLQPAKSTSHEQGKFIALFYTVVTPALNPLIYTLRNTEVKSALRHMVLENCCGSAGKLAQI
RUO
OR2C3
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates
Special Offers

Special Offers