missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human OR1F1 Full-length ORF (NP_036492.1, 1 a.a. - 312 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00004992-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16118601

  • 4115.00 NOK / 10µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms. [provided by RefSeq]

Sequence: MSGTNQSSVSEFLLLGLSRQPQQQHLLFVFFLSMYLATVLGNLLIILSVSIDSCLHTPMYFFLSNLSFVDICFSFTTVPKMLANHILETQTISFCGCLTQMYFVFMFVDMDNFLLAVMAYDHFVAVCHPLHYTAKMTHQLCALLVAGLWVVANLNVLLHTLLMAPLSFCADNAITHFFCDVTPLLKLSCSDTHLNEVIILSEGALVMITPFLCILASYMHITCTVLKVPSTKGRWKAFSTCGSHLAVVLLFYSTIIAVYFNPLSSHSAEKDTMATVLYTVVTPMLNPFIYSLRNRYLKGALKKVVGRVVFSV
Specifications

Specifications

NP_036492.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
61.3kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
OLFMF/OR16-36/OR16-37/OR16-88/OR16-89/OR16-90/OR1F10/OR1F13P/OR1F4/OR1F5/OR1F6/OR1F7/OR1F8/OR1F9/OR3-145/ORL1023
OR1F1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
4992
OR1F1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSGTNQSSVSEFLLLGLSRQPQQQHLLFVFFLSMYLATVLGNLLIILSVSIDSCLHTPMYFFLSNLSFVDICFSFTTVPKMLANHILETQTISFCGCLTQMYFVFMFVDMDNFLLAVMAYDHFVAVCHPLHYTAKMTHQLCALLVAGLWVVANLNVLLHTLLMAPLSFCADNAITHFFCDVTPLLKLSCSDTHLNEVIILSEGALVMITPFLCILASYMHITCTVLKVPSTKGRWKAFSTCGSHLAVVLLFYSTIIAVYFNPLSSHSAEKDTMATVLYTVVTPMLNPFIYSLRNRYLKGALKKVVGRVVFSV
RUO
OR1F1
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers