missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human OCRL Full-length ORF (AAH94726.1, 1 a.a. - 893 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00004952-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16138561

  • 4115.00 NOK / 10µg
Estimated Shipment: 18-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a phosphatase enzyme that is involved in actin polymerization and is found in the trans-Golgi network. Mutations in this gene cause oculocerebrorenal syndrome of Lowe and also Dent disease. [provided by RefSeq]

Sequence: MEPPLPVGAQPLATVEGMEMKGPLREPCALTLAQRNGQYELIIQLHEKEQHVQDIIPINSHFRCVQEAEETLLIDIASNSGCKIRVQGDWIRERRFEIPDEEHCLKFLSAVLAAQKAQSQLLVPEQKDSSSWYQKLDTKDKPSVFSGLLGFEDNFSSMNLDKKINSQNQPTGIHREPPPPPFSVNKMLPREKEASNKEQPKVTNTMRKLFVPNTQSGQREGLIKHILAKREKEYVNIQTFRFFVGTWNVNGQSPDSGLEPWLNCDPNPPDIYCIGFQELDLSTEAFFYFESVKEQEWSMAVERGLHSKAKYKKVQLVRLVGMMLLIFARKDQCRYIRDIATETVGTGIMGKMGNKGGVAVRFVFHNTTFCIVNSHLAAHVEDFERRNQDYKDICARMSFVVPNQTLPQLNIMKHEVVIWLGDLNYRLGMPDANEVKSLINKKDLQRLLKFDQLNIQRTQKKAFVDFNEGEIKFIPTYKYDSKTDRWDSSGKCRVPAWCDRILWRGTNVNQLNYRSHMELKTSDHKPVSALFHIGVKVVDERRYRKVFEDSVRIMDRMENDFLPSLELSRREFVFENVKFRQLQKEKFQISNNGQVPCHFSFIPKLNDSQYCKPWLRAEPFEGYLEPNETVDISLDVYVSKDSVTILNSGEDKIEDILVLHLDRGKDYFLTISGNYLPSCFGTSLEALCRMKRPIREVPVTKLIDLEKSLLQMVPLDEGASERPLQVPKEIWLLVDHLFKYACHQEDLFQTPGMQEELQQIIDCLDTSIPETIPGSNHSVAEALLIFLEALPEPVICYELYQRCLDSAYDPRICRQVISQLPRCHRNVFRYLMAFLRELLKFSEYNSVNANMIATLFTSLLLRPPPNLMARQTPSDRQRAIQFLLGFLLGSEED
Specifications

Specifications

AAH94726.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
129.6kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
INPP5F/LOCR/NPHL2/OCRL1
OCRL
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
4952
OCRL (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MEPPLPVGAQPLATVEGMEMKGPLREPCALTLAQRNGQYELIIQLHEKEQHVQDIIPINSHFRCVQEAEETLLIDIASNSGCKIRVQGDWIRERRFEIPDEEHCLKFLSAVLAAQKAQSQLLVPEQKDSSSWYQKLDTKDKPSVFSGLLGFEDNFSSMNLDKKINSQNQPTGIHREPPPPPFSVNKMLPREKEASNKEQPKVTNTMRKLFVPNTQSGQREGLIKHILAKREKEYVNIQTFRFFVGTWNVNGQSPDSGLEPWLNCDPNPPDIYCIGFQELDLSTEAFFYFESVKEQEWSMAVERGLHSKAKYKKVQLVRLVGMMLLIFARKDQCRYIRDIATETVGTGIMGKMGNKGGVAVRFVFHNTTFCIVNSHLAAHVEDFERRNQDYKDICARMSFVVPNQTLPQLNIMKHEVVIWLGDLNYRLGMPDANEVKSLINKKDLQRLLKFDQLNIQRTQKKAFVDFNEGEIKFIPTYKYDSKTDRWDSSGKCRVPAWCDRILWRGTNVNQLNYRSHMELKTSDHKPVSALFHIGVKVVDERRYRKVFEDSVRIMDRMENDFLPSLELSRREFVFENVKFRQLQKEKFQISNNGQVPCHFSFIPKLNDSQYCKPWLRAEPFEGYLEPNETVDISLDVYVSKDSVTILNSGEDKIEDILVLHLDRGKDYFLTISGNYLPSCFGTSLEALCRMKRPIREVPVTKLIDLEKSLLQMVPLDEGASERPLQVPKEIWLLVDHLFKYACHQEDLFQTPGMQEELQQIIDCLDTSIPETIPGSNHSVAEALLIFLEALPEPVICYELYQRCLDSAYDPRICRQVISQLPRCHRNVFRYLMAFLRELLKFSEYNSVNANMIATLFTSLLLRPPPNLMARQTPSDRQRAIQFLLGFLLGSEED
RUO
OCRL
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers