Learn More
Abnova™ Human NUDT5 Full-length ORF (NP_054861.2, 1 a.a. - 219 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00011164-P01.25ug
Additional Details : Weight : 0.00010kg
Description
Nudix hydrolases, such as NUDT5, eliminate toxic nucleotide derivatives from the cell and regulate the levels of important signaling nucleotides and their metabolites (McLennan, 1999 [PubMed 10373642]).[supplied by OMIM]
Sequence: MESQEPTESSQNGKQYIISEELISEGKWVKLEKTTYMDPTGKTRTWESVKRTTRKEQTADGVAVIPVLQRTLHYECIVLVKQFRPPMGGYCIEFPAGLIDDGETPEAAALRELEEETGYKGDIAECSPAVCMDPGLSNCTIHIVTVTINGDDAENARPKPKPGDGEFVEVISLPKNDLLQRLDALVAEEHLTVDARVYSYALALKHANAKPFEVPFLKFSpecifications
NP_054861.2 | |
Liquid | |
11164 | |
NUDT5 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MESQEPTESSQNGKQYIISEELISEGKWVKLEKTTYMDPTGKTRTWESVKRTTRKEQTADGVAVIPVLQRTLHYECIVLVKQFRPPMGGYCIEFPAGLIDDGETPEAAALRELEEETGYKGDIAECSPAVCMDPGLSNCTIHIVTVTINGDDAENARPKPKPGDGEFVEVISLPKNDLLQRLDALVAEEHLTVDARVYSYALALKHANAKPFEVPFLKF | |
RUO | |
NUDT5 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
50.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
YSA1/YSA1H/hYSAH1 | |
NUDT5 | |
Yes | |
wheat germ expression system |