missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NUBP1 Full-length ORF (NP_002475.2, 1 a.a. - 320 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00004682-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16148161

  • 6245.00 NOK / 25µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

NUBP1 is a member of the NUBP/MRP subfamily of ATP-binding proteins (Nakashima et al., 1999 [PubMed 10486206]).[supplied by OMIM]

Sequence: MEEVPHDCPGADSAQAGRGASCQGCPNQRLCASGAGATPDTAIEEIKEKMKTVKHKILVLSGKGGVGKSTFSAHLAHGLAEDENTQIALLDIDICGPSIPKIMGLEGEQVHQSGSGWSPVYVEDNLGVMSVGFLLSSPDDAVIWRGPKKNGMIKQFLRDVDWGEVDYLIVDTPPGTSDEHLSVVRYLATAHIDGAVIITTPQEVSLQDVRKEINFCRKVKLPIIGVVENMSGFICPKCKKESQIFPPTTGGAELMCQDLEVPLLGRVPLDPLIGKNCDKGQSFFIDAPDSPATLAYRSIIQRIQEFCNLHQSKEENLISS
Specifications

Specifications

NP_002475.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
60.9kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC117406/MGC130052/MGC130053/NBP/NBP1
NUBP1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
4682
NUBP1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MEEVPHDCPGADSAQAGRGASCQGCPNQRLCASGAGATPDTAIEEIKEKMKTVKHKILVLSGKGGVGKSTFSAHLAHGLAEDENTQIALLDIDICGPSIPKIMGLEGEQVHQSGSGWSPVYVEDNLGVMSVGFLLSSPDDAVIWRGPKKNGMIKQFLRDVDWGEVDYLIVDTPPGTSDEHLSVVRYLATAHIDGAVIITTPQEVSLQDVRKEINFCRKVKLPIIGVVENMSGFICPKCKKESQIFPPTTGGAELMCQDLEVPLLGRVPLDPLIGKNCDKGQSFFIDAPDSPATLAYRSIIQRIQEFCNLHQSKEENLISS
RUO
NUBP1
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers